Please login to your online account to display your discounted pricing

Alfa Aesar™ Isopropyl 3-acetylpyridine-2-carboxylate, 97%

$90.50 - $90.50

Chemical Identifiers

Chemical Name or Material Isopropyl 3-acetylpyridine-2-carboxylate
CAS 195812-68-1
Molecular Formula C11H13NO3
Formula Weight 207.23
MDL Number MFCD01314548
Beilstein 7760203
Synonym 3-Acetylpyridine-2-carboxylic acid isopropyl ester; Isopropyl 3-acetylpicolinate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1358306

 
alfa aesar™
L1358306
5g Each for $90.50
Description & Specifications

Specifications

Formula Weight 207.23
Chemical Name or Material Isopropyl 3-acetylpyridine-2-carboxylate
Assay 97%
Molecular Formula C11H13NO3
MDL Number MFCD01314548
Beilstein 7760203
CAS 195812-68-1
Synonym 3-Acetylpyridine-2-carboxylic acid isopropyl ester; Isopropyl 3-acetylpicolinate
Boiling Point 140° to 142°C (1mmHg)
Melting Point 38° to 42°C
Quantity 5g