missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropyl 2-Methylbutyrate, TCI America™

$45.83 - $224.59

Chemical Identifiers

CAS 66576-71-4
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00085203
InChI Key DIRDKDDFAMNBNY-UHFFFAOYSA-N
Synonym 2-Methylbutyric Acid Isopropyl Ester
PubChem CID 522214
IUPAC Name propan-2-yl 2-methylbutanoate
SMILES CCC(C)C(=O)OC(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I073625ML View Documents TCI America
I073625ML
25mL Each for $45.83
 
I0736500ML View Documents TCI America
I0736500ML
500mL Each for $224.59
 
Specifications

Chemical Identifiers

66576-71-4
144.214
DIRDKDDFAMNBNY-UHFFFAOYSA-N
522214
CCC(C)C(=O)OC(C)C
C8H16O2
MFCD00085203
2-Methylbutyric Acid Isopropyl Ester
propan-2-yl 2-methylbutanoate

Specifications

144°C
25mL
C8H16O2
3272
DIRDKDDFAMNBNY-UHFFFAOYSA-N
propan-2-yl 2-methylbutanoate
522214
Clear Liquid at 20°C
Isopropyl 2-Methylbutyrate
Colorless
66576-71-4
MFCD00085203
2-Methylbutyric Acid Isopropyl Ester
CCC(C)C(=O)OC(C)C
144.214
144.21
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : Yes