missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isopropenyl chloroformate, 96%, stabilized, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC275120010
Description
Coupling reagentSpecifications
Isopropenyl chloroformate | |
10°C | |
Glass bottle | |
1.103 | |
95.0°C to 97.0°C | |
1g | |
57933-83-2 | |
C4H5ClO2 | |
MFCD00037083 | |
08,53 | |
Solubility in water: reacts with water | |
CC(=C)OC(=O)Cl | |
120.54 | |
120.54 | |
96% |
1.1030g/mL | |
Authentic | |
1.4130 to 1.4150 | |
50 to 100ppm BHT | |
Colorless | |
Stabilized | |
95% min. (GC) | |
CH2=C(CH3)CO2Cl | |
03,28 | |
carbonochloridic acid, 1-methylethenyl ester, chloro prop-1-en-2-yloxy methanone, chloroformic acid isopropenyl ester, chloroformic acid,isopropenyl ester, isopropenyl carbonochloridate, isopropenyl chloridocarbonate, isopropenyl chloroformate, isopropenylchloro-formate, isopropenylchloroformate, prop-1-en-2-yl chloroformate | |
AMMGCGVWJMRTQI-UHFFFAOYSA-N | |
prop-1-en-2-yl carbonochloridate | |
93846 | |
Liquid |
Chemical Identifiers
57933-83-2 | |
120.54 | |
AMMGCGVWJMRTQI-UHFFFAOYSA-N | |
93846 | |
CC(=C)OC(=O)Cl |
C4H5ClO2 | |
MFCD00037083 | |
carbonochloridic acid, 1-methylethenyl ester, chloro prop-1-en-2-yloxy methanone, chloroformic acid isopropenyl ester, chloroformic acid,isopropenyl ester, isopropenyl carbonochloridate, isopropenyl chloridocarbonate, isopropenyl chloroformate, isopropenylchloro-formate, isopropenylchloroformate, prop-1-en-2-yl chloroformate | |
prop-1-en-2-yl carbonochloridate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse
GHS Signal Word: Danger
EINECSNumber : 261-018-4
Recommended Storage : Product may turn yellow during storage