Please login to your online account to display your discounted pricing

Isopropenyl chloroformate, 97%, stabilized, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  275120050

 View more versions of this product

Catalog No. AC275120050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Quantity 5g
Color Clear colorless
CAS 57933-83-2
MDL Number MFCD00037083
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.
Chemical Name or Material Isopropenyl chloroformate
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note Stabilized
Storage Information Product may turn yellow during storage
Molecular Formula C4H5ClO2
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4130 to 1.4150 (20°C, 589nm)
Stabilizer 50 to 100ppm BHT
Health Hazard 1 Danger
Linear Formula CH2=C(CH3)CO2Cl
Assay Percent Range 97%
Density 1.1030g/mL
Formula Weight 120.54

Specifications

Chemical Name or Material Isopropenyl chloroformate
Name Note Stabilized
CAS 57933-83-2
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H5ClO2
Linear Formula CH2=C(CH3)CO2Cl
Formula Weight 120.54
MDL Number MFCD00037083
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.

SDS & Safety Documents