missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Isopropenyl acetate, 99%

Manufacturer:  Alfa Aesar™ A15618AE

 View more versions of this product

Catalog No. AAA15618AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Applications
Iodine in isopropenyl acetate is a unique catalyst for the acetylation of a variety of alcohols, phenols and amines under solvent free conditions. It is used as a solvent for cellulose, plastics, oil and fats. It is also a building block used for various organic synthesis.

Solubility
Slightly soluble in water. Soluble in alcohol and propylene glycol, acetone and ethyl ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Isopropenyl acetate
93°C to 94°C
100mL
C5H8O2
UN2403
14,5203
HETCEOQFVDFGSY-UHFFFAOYSA-N
prop-1-en-2-yl acetate
7916
99%
18°C (64°F)
-93°C
108-22-5
MFCD00008709
1280347
isopropenyl acetate, 2-acetoxypropene, 1-methylvinyl acetate, 2-acetoxypropylene, methylvinyl acetate, 1-propen-2-ol, acetate, propen-2-yl acetate, acetic acid isopropenyl ester, 1-acetoxy-1-methylethylene, 1-propen-2-yl acetate
CC(=C)OC(=O)C
100.117
100.12

Chemical Identifiers

108-22-5
100.117
HETCEOQFVDFGSY-UHFFFAOYSA-N
7916
CC(=C)OC(=O)C
C5H8O2
MFCD00008709
isopropenyl acetate, 2-acetoxypropene, 1-methylvinyl acetate, 2-acetoxypropylene, methylvinyl acetate, 1-propen-2-ol, acetate, propen-2-yl acetate, acetic acid isopropenyl ester, 1-acetoxy-1-methylethylene, 1-propen-2-yl acetate
prop-1-en-2-yl acetate
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPENYL ACETATE

RTECSNumber : UD4200000

TSCA : TSCA

Documents