Learn More
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
$38.53 - $128.50
Chemical Identifiers
CAS | 67-63-0 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.096 |
MDL Number | MFCD00011674 |
InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
PubChem CID | 3776 |
ChEBI | CHEBI:17824 |
IUPAC Name | propan-2-ol |
SMILES | CC(C)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC396982500
|
Thermo Scientific Chemicals
396982500 |
250 mL |
Each for $38.53
|
|
|||||
AC396980010
|
Thermo Scientific Chemicals
396980010 |
1 L |
Each for $95.02
|
|
|||||
AC396980025
|
Thermo Scientific Chemicals
396980025 |
2.5 L |
Each for $128.50
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
67-63-0 | |
60.096 | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
C3H8O | |
MFCD00011674 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
Specifications
-89.5°C | |
0.01 max. (EurPh) at 310nm, 0.02 max. (EurPh) at 290nm, 0.03 max. (EurPh) at 270nm, 0.1 max. (EurPh) at 250nm, 0.3 max. (EurPh) at 230nm | |
Clear colorless liquid | |
81.0°C to 83.0°C | |
12°C | |
Authentic | |
Low Melting Solid | |
0.785 | |
(CH3)2CHOH | |
2.27 mPa.s (20°C) | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.1 | |
Isopropanol |
67-63-0 | |
Brown to Purple | |
0.7850g/mL | |
250 mL | |
C: Passes Test (EP) | |
Glass Bottle | |
1.3760 to 1.3780 | |
C3H8O | |
MFCD00011674 | |
0.5% max. | |
Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, methylene chloride,, dmf and acetone, miscible with alcohol, ether and chloroform, insoluble in salts solutions | |
CC(C)O | |
60.096 | |
CHEBI:17824 | |
≥99% |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness
Causes serious eye irritation
Highly flammable liquid and vapor
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray
Wear protective gloves/ protective clothing/face protection
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing
Keep away from heat/sparks/open flames/hot surfaces. - No smoking
Ground/Bond container and receiving equipment
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 200-661-7
RUO – Research Use Only