Please login to your online account to display your discounted pricing

Isopropanol-d8, for NMR, +99% atom D, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  174850250

 View more versions of this product

Catalog No. AC174850250

  • / Each
 

Description & Specifications

Chemical Identifiers

Deuterium (D) 99.0% min.
Packaging Glass bottle
Density 0.8900g/mL
Quantity 25g
Color Clear colorless
Chemical Name or Material Isopropanol-d8
Name Note 99+ atom % D
CAS 22739-76-0
Purity Grade For NMR
Physical Form Clear colorless liquid
Molecular Formula C3D8O
Linear Formula (CD3)2CDOD
Formula Weight 68.15g/mol
MDL Number MFCD00044341
Synonym 2-Propanol-d8
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 245-189-2

Specifications

Chemical Name or Material Isopropanol-d8
Name Note 99+ atom % D
CAS 22739-76-0
Purity Grade For NMR
Molecular Formula C3D8O
Linear Formula (CD3)2CDOD
Formula Weight 68.15g/mol
MDL Number MFCD00044341
Synonym 2-Propanol-d8

For NMR

Accessories

Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents