Please login to your online account to display your discounted pricing

Isopropanol-d8, for NMR, +99% atom D, ACROS Organics™

$114.19 - $387.38

Chemical Identifiers

Chemical Name or Material Isopropanol-d8
Name Note 99+ atom % D
CAS 22739-76-0
Purity Grade For NMR
Molecular Formula C3D8O
Linear Formula (CD3)2CDOD
Formula Weight 68.15
MDL Number MFCD00044341
Synonym 2-Propanol-d8
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC174850050

  SDS
thermo fisher scientific
174850050
Glass bottle 5g Each for $114.19

AC174850250

  SDS
thermo fisher scientific
174850250
Glass bottle 25g Each for $387.38
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 68.15
Quantity 5g
Color Clear colorless
EINECS Number 245-189-2
Purity Grade For NMR
Density 0.8900g/mL
Deuterium (D) 99.0% min.
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
MDL Number MFCD00044341
Health Hazard 1 Danger
Synonym 2-Propanol-d8
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Isopropanol-d8
Name Note 99+ atom % D
Linear Formula (CD3)2CDOD
CAS 22739-76-0
Molecular Formula C3D8O

For NMR

Safety and Handling

Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.