Please login to your online account to display your discounted pricing

Isopropanol, 99.8%, for electronic use, residue free, ACROS Organics™


Quantity 2.5L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC327930025

 View more versions of this product

Catalog No. AC327930025

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.8%
Physical Form Liquid
Formula Weight 60.1
Quantity 2.5L
Color Clear
Purity Grade For electronic use
Density 0.7850g/mL
Beilstein 01, 360
Merck Index 15, 5254
Fieser 13,108
Arsenic (As) 10ppb max.
Packaging Glass bottle
Linear Formula (CH3)2CHOH
Chlorine (Cl) 2ppm max.
Copper (Cu) 10ppb max.
Name Note Residue Free
Molecular Formula C3H8O
MDL Number MFCD00011674
CAS 67-63-0
Chemical Name or Material Isopropanol
Assay Percent Range 99.8%
Health Hazard 1 Danger
Barium (Ba) 20ppb max.
Acidity 0.01% max.
Iron (Fe) 50ppb max.
Residue after Evaporation 0.0003% max.
Synonym 2-Propanol
Water 0.05% max. (Coulometric)
Heavy Metals (as Pb) 1ppm max.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.

Specifications

Chemical Name or Material Isopropanol
Name Note Residue Free
CAS 67-63-0
Purity Grade For electronic use
Assay ≥99.8%
Assay Percent Range 99.8%
Molecular Formula C3H8O
Linear Formula (CH3)2CHOH
Formula Weight 60.1
MDL Number MFCD00011674
Synonym 2-Propanol
Accessories

Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents