Please login to your online account to display your discounted pricing

Isopropanol, 99.6%, ACS reagent, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: fisher scientific acros  423835000

 View more versions of this product

Catalog No. AC423835000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.5%
Formula Weight 60.1
Physical Form Liquid
Purity Grade ACS Reagent Grade
Quantity 500mL
Color Clear
Beilstein 01, 360
Fieser 13,108
Merck Index 15, 5254
Solubility Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, methylene chloride, dmf and acetone, miscible with alcohol, ether and chloroform, insoluble in salts solutions
Density 0.7850g/mL
Residue after Evaporation 0.001% max.
Packaging Glass bottle
Molecular Formula C3H8O
MDL Number MFCD00011674
CAS 67-63-0
Assay Percent Range 99.6%
Chemical Name or Material Isopropanol
Synonym 2-Propanol
Carbonyl Compounds 0.002% max. propionaldehyde or acetone
Additional Information Vapor Pressure: 43mmHg at 20°C
Water 0.2% max. (K.F.)
Linear Formula (CH3)2CHOH
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Isopropanol
CAS 67-63-0
Purity Grade ACS Reagent Grade
Assay ≥99.5%
Assay Percent Range 99.6%
Molecular Formula C3H8O
Linear Formula (CH3)2CHOH
Formula Weight 60.1
MDL Number MFCD00011674
Synonym 2-Propanol
Accessories

Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents