missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropanol, 99.6%, ACS reagent, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 423830200

 View more versions of this product

Catalog No. AC423830200


 

Specifications Description & Specifications

Specifications

≥99.5%
ACS Reagent Grade
20L
15, 5254
13,108
0.7850g/mL
(CH3)2CHOH
Isopropanol
0.001% max.
0.002% max. propionaldehyde or acetone
C3H8O
Liquid
60.1
Clear
01, 360
Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, methylene chloride, dmf and acetone, miscible with alcohol, ether and chloroform, insoluble in salts solutions
67-63-0
0.2% max. (K.F.)
MFCD00011674
2-Propanol
Vapor Pressure: 43mmHg at 20°C
99.6%

Chemical Identifiers

67-63-0
C3H8O
60.1
2-Propanol
≥99.5%
99.6%
(CH3)2CHOH
MFCD00011674
Isopropanol
ACS Reagent Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger