Please login to your online account to display your discounted pricing

Isopropanol, 99.5%, for spectroscopy ACS, ACROS Organics™

$47.83 - $138.08

Chemical Identifiers

Chemical Name or Material Isopropanol
CAS 67-63-0
Purity Grade For spectroscopy ACS
Assay ≥99.5%
Assay Percent Range 99.5%
Molecular Formula C3H8O
Linear Formula (CH3)2CHOH
Formula Weight 60.1
MDL Number MFCD00011674
Synonym 2-Propanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC412790010

  SDS
thermo fisher scientific
AC412790010
Glass bottle 1L Each for $79.78

AC412790025

  SDS
thermo fisher scientific
AC412790025
Glass bottle 2.5L Each for $138.08

AC412795000

  SDS
thermo fisher scientific
412795000
EcoSafPak™ Glass Bottle 500mL Each for $47.83
Description & Specifications

Specifications

Assay ≥99.5%
Physical Form Liquid
Formula Weight 60.1
Quantity 1L
Color Clear
Beilstein 01, 360
Fieser 13,108
Merck Index 15, 5254
Solubility Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, methylene chloride, dmf and acetone, miscible with alcohol, ether and chloroform, insoluble in salts solutions
TSCA TSCA
EINECS Number 200-661-7
RTECS Number NT8050000
Purity Grade For spectroscopy ACS
Density 0.7850g/mL
Residue after Evaporation 0.001% max.
Absorbance 0.01 max. at 330nm
Water 0.2% max. (K.F.)
Synonym 2-Propanol
CAS 67-63-0
Molecular Formula C3H8O
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Assay Percent Range 99.5%
MDL Number MFCD00011674
Additional Information Vapor Pressure: 43mmHg at 20°C
Carbonyl Compounds 0.002% max. propionaldehyde or acetone
Packaging Glass bottle
Linear Formula (CH3)2CHOH
Health Hazard 1 Danger
Chemical Name or Material Isopropanol
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

For UV/VIS spectroscopy

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.