Please login to your online account to display your discounted pricing

Isopropanol, 99.5%, for spectroscopy ACS, ACROS Organics™


Quantity 500mL
Packaging EcoSafPak™ Glass Bottle

Manufacturer: thermo fisher scientific  412795000

 View more versions of this product

Catalog No. AC412795000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.5%
Boiling Point 81.0° to 83.0°C
Formula Weight 60.1
Physical Form Liquid
Quantity 500mL
Color Clear
Packaging EcoSafPak™ Glass Bottle
TSCA TSCA
EINECS Number 200-661-7
Solubility Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, methylene chloride, dmf and acetone, miscible with alcohol, ether and chloroform, insoluble in salts solutions
Purity Grade For spectroscopy ACS
RTECS Number NT8050000
Density 0.7850g/mL
Beilstein 01, 360
Merck Index 15, 5254
Fieser 13,108
Residue after Evaporation 0.001% max.
Absorbance 0.01 max. at 330nm
Synonym 2-Propanol
Carbonyl Compounds 0.002% max. propionaldehyde or acetone
Water 0.2% max. (K.F.)
CAS 67-63-0
Chemical Name or Material Isopropanol
Assay Percent Range 99.5%
Molecular Formula C3H8O
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00011674
Additional Information Vapor Pressure: 43mmHg at 20°C
Health Hazard 1 Danger
Linear Formula (CH3)2CHOH
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.

Specifications

Chemical Name or Material Isopropanol
CAS 67-63-0
Purity Grade For spectroscopy ACS
Assay ≥99.5%
Assay Percent Range 99.5%
Molecular Formula C3H8O
Linear Formula (CH3)2CHOH
Formula Weight 60.1
MDL Number MFCD00011674
Synonym 2-Propanol
Accessories

Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents