missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Isoprene monoxide, 97%

Manufacturer:  Alfa Aesar™ L1976806

 View more versions of this product

Catalog No. AAL1976806



Description

Description

Applications
Isoprene monoxide is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. Recommended storage temperature is 2 - 8°C.
Specifications

Specifications

Isoprene monoxide
Recognizable
5g
C5H8O
UN1993
2-methyl-2-vinyloxirane, isoprene monoxide, ccris 5685, 3,4-epoxy-3-methyl-1-butene, 2-methyl-2-vinyl-oxirane, oxirane, 2-ethenyl-2-methyl-, 2r, acmc-20m5m5, oxirane, 2-ethenyl-2-methyl, 2-methyl-2-vinyloxirane, 4,4,4-3fmet-1-2-furyl-1,3-butandione
CC1(CO1)C=C
84.118
84.12
−8°C (17°F)
79°C to 81°C
1838-94-4
MFCD00075225
104216
FVCDMHWSPLRYAB-UHFFFAOYSA-N
2-ethenyl-2-methyloxirane
92166
97%

Chemical Identifiers

1838-94-4
84.118
FVCDMHWSPLRYAB-UHFFFAOYSA-N
92166
CC1(CO1)C=C
C5H8O
MFCD00075225
2-methyl-2-vinyloxirane, isoprene monoxide, ccris 5685, 3,4-epoxy-3-methyl-1-butene, 2-methyl-2-vinyl-oxirane, oxirane, 2-ethenyl-2-methyl-, 2r, acmc-20m5m5, oxirane, 2-ethenyl-2-methyl, 2-methyl-2-vinyloxirane, 4,4,4-3fmet-1-2-furyl-1,3-butandione
2-ethenyl-2-methyloxirane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280g-P243-P305+P351+P338-P403
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
Take precautionary measures against static discharge.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Recommended Storage : 2° to 8°C

Documents