Learn More
Isoprene monoxide, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1976814
Description
Isoprene monoxide is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsoprene monoxide is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. Recommended storage temperature is 2 - 8°C.
Specifications
Isoprene monoxide | |
0.857 | |
−8°C (17°F) | |
C5H8O | |
MFCD00075225 | |
UN1993 | |
2-methyl-2-vinyloxirane, isoprene monoxide, ccris 5685, 3,4-epoxy-3-methyl-1-butene, 2-methyl-2-vinyl-oxirane, oxirane, 2-ethenyl-2-methyl-, 2r, acmc-20m5m5, oxirane, 2-ethenyl-2-methyl, 4,4,4-3fmet-1-2-furyl-1,3-butandione | |
FVCDMHWSPLRYAB-UHFFFAOYSA-N | |
2-ethenyl-2-methyloxirane | |
92166 | |
97% |
1838-94-4 | |
79°C to 81°C | |
Recognizable | |
1.416 | |
25 g | |
104216 | |
Slightly soluble in water. | |
CC1(CO1)C=C | |
84.118 | |
84.12 |
Chemical Identifiers
1838-94-4 | |
84.118 | |
FVCDMHWSPLRYAB-UHFFFAOYSA-N | |
92166 | |
CC1(CO1)C=C |
C5H8O | |
MFCD00075225 | |
2-methyl-2-vinyloxirane, isoprene monoxide, ccris 5685, 3,4-epoxy-3-methyl-1-butene, 2-methyl-2-vinyl-oxirane, oxirane, 2-ethenyl-2-methyl-, 2r, acmc-20m5m5, oxirane, 2-ethenyl-2-methyl, 4,4,4-3fmet-1-2-furyl-1,3-butandione | |
2-ethenyl-2-methyloxirane |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only