missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Isopore™ Membrane Filters, 0.6 μm

Catalog No. DTTP04700
Change view
Click to view available options
Diameter (Metric):
13 mm
25 mm
47 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
DTTP04700 47 mm
DTTP01300 13 mm
DTTP02500 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. DTTP04700 Supplier MilliporeSigma™ Supplier No. DTTP04700
Only null left

Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.

The Isopore™ membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore™ membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore™ membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.

Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety

Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays

Specifications

Autoclavable Not Autoclavable
Bubble Point 8.5 psi
Color White
Filter Type Screen Filter
Material Polycarbonate
Pore Size 0.6 μm
Sterility Non-sterile
Flow Rate Avg. 43 mL/min x cm2 (typical results (at 10 psi);62 mL/min x cm2/psi)
For Use With (Application) Air Analysis; Cell Biology; Cell Culture; Bioassays; Cytology; Environmental Analysis; Epifluorescence.
Gas Flow Rate 10.0 L/min/cm2
Hydrophobicity Hydrophilic
No. per Case 100Pack
Quantity 100/Pk.
Type Filter Membranes
Wettability Hydrophilic
Diameter (Metric) 47 mm
Show More Show Less