Learn More
Isophthalic acid, 99%, Thermo Scientific Chemicals
$39.62 - $187.49
Chemical Identifiers
CAS | 121-91-5 |
---|---|
Molecular Formula | C8H6O4 |
Molecular Weight (g/mol) | 166.132 |
MDL Number | MFCD00002516 |
InChI Key | QQVIHTHCMHWDBS-UHFFFAOYSA-N |
Synonym | isophthalic acid, 1,3-benzenedicarboxylic acid, m-phthalic acid, m-benzenedicarboxylic acid, acide isophtalique, kyselina isoftalova, iso-phthalic acid, acide isophtalique french, kyselina isoftalova czech, unii-x35216h9fj |
PubChem CID | 8496 |
ChEBI | CHEBI:30802 |
IUPAC Name | benzene-1,3-dicarboxylic acid |
SMILES | C1=CC(=CC(=C1)C(=O)O)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1444530
|
Thermo Scientific Chemicals
A1444530 |
250 g |
Each for $39.62
|
|
|||||
AAA144450B
|
Thermo Scientific Chemicals
A144450B |
1000 g |
Each for $53.44
|
|
|||||
AAA144450I
|
Thermo Scientific Chemicals
A144450I |
5000 g |
Each for $187.49
|
|
|||||
Description
Isophthalic acid is used as an intermediate for high performance unsaturated polyesters, resins for coatings, high solids paints, gel coats and modifier of polyethylene terephthalate for bottles. It acts as precursors for the fire-resistant material nomex as well as used in the preparation of high-performance polymer polybenzimidazole. It is also employed as an input for the production of insulation materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsophthalic acid is used as an intermediate for high performance unsaturated polyesters, resins for coatings, high solids paints, gel coats and modifier of polyethylene terephthalate for bottles. It acts as precursors for the fire-resistant material nomex as well as used in the preparation of high-performance polymer polybenzimidazole. It is also employed as an input for the production of insulation materials.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
121-91-5 | |
166.132 | |
QQVIHTHCMHWDBS-UHFFFAOYSA-N | |
8496 | |
benzene-1,3-dicarboxylic acid |
C8H6O4 | |
MFCD00002516 | |
isophthalic acid, 1,3-benzenedicarboxylic acid, m-phthalic acid, m-benzenedicarboxylic acid, acide isophtalique, kyselina isoftalova, iso-phthalic acid, acide isophtalique french, kyselina isoftalova czech, unii-x35216h9fj | |
CHEBI:30802 | |
C1=CC(=CC(=C1)C(=O)O)C(=O)O |
Specifications
121-91-5 | |
1.54 | |
MFCD00002516 | |
1909332 | |
isophthalic acid, 1,3-benzenedicarboxylic acid, m-phthalic acid, m-benzenedicarboxylic acid, acide isophtalique, kyselina isoftalova, iso-phthalic acid, acide isophtalique french, kyselina isoftalova czech, unii-x35216h9fj | |
QQVIHTHCMHWDBS-UHFFFAOYSA-N | |
benzene-1,3-dicarboxylic acid | |
8496 | |
166.13 | |
Isophthalic acid |
344°C to 348°C (sublimation) | |
C8H6O4 | |
250 g | |
14,5197 | |
Insoluble in water. | |
C1=CC(=CC(=C1)C(=O)O)C(=O)O | |
166.132 | |
CHEBI:30802 | |
99% |
Safety and Handling
EINECSNumber : 204-506-4
RTECSNumber : NT2007000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only