missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isophthalaldehydic Acid, TCI America™

$76.39 - $226.11

Chemical Identifiers

CAS 619-21-6
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.133
MDL Number MFCD00039575
InChI Key UHDNUPHSDMOGCR-UHFFFAOYSA-N
Synonym 3-carboxybenzaldehyde, benzoic acid, 3-formyl, m-formyl benzoic acid, 3-formyl benzoic acid, 3-formyl-benzoic acid, unii-hb95grm95d, m-carboxybenzaldehyde, hb95grm95d, 3-formylbenzoicacid, isophthalaldehydsaure
PubChem CID 12077
IUPAC Name 3-formylbenzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I08515G View Documents TCI America
I08515G
5g Each for $226.11
 
I08511G View Documents TCI America
I08511G
1g Each for $76.39
 
Specifications

Chemical Identifiers

619-21-6
150.133
UHDNUPHSDMOGCR-UHFFFAOYSA-N
12077
C1=CC(=CC(=C1)C(=O)O)C=O
C8H6O3
MFCD00039575
3-carboxybenzaldehyde, benzoic acid, 3-formyl, m-formyl benzoic acid, 3-formyl benzoic acid, 3-formyl-benzoic acid, unii-hb95grm95d, m-carboxybenzaldehyde, hb95grm95d, 3-formylbenzoicacid, isophthalaldehydsaure
3-formylbenzoic acid

Specifications

White-Yellow
5g
C8H6O3
3-carboxybenzaldehyde, benzoic acid, 3-formyl, m-formyl benzoic acid, 3-formyl benzoic acid, 3-formyl-benzoic acid, unii-hb95grm95d, m-carboxybenzaldehyde, hb95grm95d, 3-formylbenzoicacid, isophthalaldehydsaure
C1=CC(=CC(=C1)C(=O)O)C=O
150.133
150.13
≥98.0% (HPLC,T)
177°C
619-21-6
MFCD00039575
UHDNUPHSDMOGCR-UHFFFAOYSA-N
3-formylbenzoic acid
12077
Crystal-Powder at 20°C
Isophthalaldehydic Acid
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1670

TSCA : Yes