missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isophthalaldehyde, 99%, Thermo Scientific™
Chemical Identifiers
CAS | 626-19-7 |
---|---|
Molecular Formula | C8H6O2 |
Molecular Weight (g/mol) | 134.13 |
MDL Number | MFCD00003372 |
InChI Key | IZALUMVGBVKPJD-UHFFFAOYSA-N |
Synonym | isophthalaldehyde, m-phthalaldehyde, 1,3-benzenedicarboxaldehyde, 1,3-benzenedialdehyde, isophtaldehydes, isophthaldehyde, isophthaldialdehyde, isophtaldehydes french, isophthalic dicarboxaldehyde, unii-lu162b2n9x |
PubChem CID | 34777 |
IUPAC Name | benzene-1,3-dicarbaldehyde |
SMILES | C1=CC(=CC(=C1)C=O)C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC151740050
|
Thermo Scientific Chemicals
151740050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC151740250
|
Thermo Scientific Chemicals
151740250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
626-19-7 | |
134.13 | |
IZALUMVGBVKPJD-UHFFFAOYSA-N | |
34777 | |
C1=CC(=CC(=C1)C=O)C=O |
C8H6O2 | |
MFCD00003372 | |
isophthalaldehyde, m-phthalaldehyde, 1,3-benzenedicarboxaldehyde, 1,3-benzenedialdehyde, isophtaldehydes, isophthaldehyde, isophthaldialdehyde, isophtaldehydes french, isophthalic dicarboxaldehyde, unii-lu162b2n9x | |
benzene-1,3-dicarbaldehyde |
Specifications
626-19-7 | |
Authentic | |
Glass bottle | |
C6H4(CHO)2 | |
5g | |
isophthalaldehyde, m-phthalaldehyde, 1,3-benzenedicarboxaldehyde, 1,3-benzenedialdehyde, isophtaldehydes, isophthaldehyde, isophthaldialdehyde, isophtaldehydes french, isophthalic dicarboxaldehyde, unii-lu162b2n9x | |
IZALUMVGBVKPJD-UHFFFAOYSA-N | |
benzene-1,3-dicarbaldehyde | |
34777 | |
99% |
136°C (17.0 hPa) | |
98.5% min. (GC) | |
C8H6O2 | |
MFCD00003372 | |
07, 675 | |
Solubility in water: slightly soluble | |
C1=CC(=CC(=C1)C=O)C=O | |
134.13 | |
134.13 | |
Isophthalaldehyde, 99% |
Safety and Handling
EINECSNumber : 210-935-8
RTECSNumber : NT1981000
RUO â Research Use Only