missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isophthalaldehyde, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 151740250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Isophthalaldehyde | |
626-19-7 | |
Authentic | |
Glass bottle | |
C6H4(CHO)2 | |
25g | |
isophthalaldehyde, m-phthalaldehyde, 1,3-benzenedicarboxaldehyde, 1,3-benzenedialdehyde, isophtaldehydes, isophthaldehyde, isophthaldialdehyde, isophtaldehydes french, isophthalic dicarboxaldehyde, unii-lu162b2n9x | |
IZALUMVGBVKPJD-UHFFFAOYSA-N | |
benzene-1,3-dicarbaldehyde | |
34777 | |
99% |
99% | |
136°C (17.0 hPa) | |
98.5% min. (GC) | |
C8H6O2 | |
MFCD00003372 | |
07, 675 | |
Solubility in water: slightly soluble | |
C1=CC(=CC(=C1)C=O)C=O | |
134.13 | |
134.13 |
Chemical Identifiers
626-19-7 | |
134.13 | |
IZALUMVGBVKPJD-UHFFFAOYSA-N | |
34777 | |
C1=CC(=CC(=C1)C=O)C=O |
C8H6O2 | |
MFCD00003372 | |
isophthalaldehyde, m-phthalaldehyde, 1,3-benzenedicarboxaldehyde, 1,3-benzenedialdehyde, isophtaldehydes, isophthaldehyde, isophthaldialdehyde, isophtaldehydes french, isophthalic dicarboxaldehyde, unii-lu162b2n9x | |
benzene-1,3-dicarbaldehyde |
Safety and Handling
EINECSNumber : 210-935-8
RTECSNumber : NT1981000
RUO â Research Use Only