Learn More
Isophorone diisocyanate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 427602500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Isophorone diisocyanate | |
Colorless to Yellow | |
153.0°C (10.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4830 to 1.4850 (20°C, 589nm) | |
250 mL | |
isophorone diisocyanate, ipdi, isophorone diamine diisocyanate, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanate, cyclohexane, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethyl, 3-isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate, ccris 6252, isocyanic acid, methylene 3,5,5-trimethyl-3,1-cyclohexylene ester, isophorone diisocyanate diisocyanates | |
CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O | |
222.29 | |
13-15 mPa.s (23°C) | |
222.28 | |
Liquid |
4098-71-9 | |
1.0610g/mL | |
163°C | |
97.5% min. (sum of mixed isomers) (GC) | |
C12H18N2O2 | |
MFCD00064956 | |
1.061 | |
NIMLQBUJDJZYEJ-UHFFFAOYNA-N | |
5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane | |
169132 | |
CHEBI:53214 | |
98% |
Chemical Identifiers
4098-71-9 | |
222.29 | |
NIMLQBUJDJZYEJ-UHFFFAOYNA-N | |
169132 | |
5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane |
C12H18N2O2 | |
MFCD00064956 | |
isophorone diisocyanate, ipdi, isophorone diamine diisocyanate, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanate, cyclohexane, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethyl, 3-isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate, ccris 6252, isocyanic acid, methylene 3,5,5-trimethyl-3,1-cyclohexylene ester, isophorone diisocyanate diisocyanates | |
CHEBI:53214 | |
CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic to aquatic life with long lasti
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If skin irritation or
GHS Signal Word: Danger
EINECSNumber : 223-861-6
RUO – Research Use Only