missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isophorone, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC122640025
Specifications
Isophorone | |
84°C | |
Glass bottle | |
0.92 | |
213.0°C to 214.0°C | |
-8.0°C | |
98% | |
96% min. (GC) | |
MFCD00001584 | |
15,5242 | |
Solubility in water: 12g/L (20°C). Other solubilities: soluble in alcohol,ether,acetone | |
CC1=CC(=O)CC(C1)(C)C | |
138.21 | |
CHEBI:34800 | |
Liquid |
0.9200g/mL | |
Authentic | |
1.4770 to 1.4790 | |
2.6 mPa.s (20°C) | |
Colorless to Yellow | |
2.5L | |
78-59-1 | |
C9H14O | |
07,65 | |
1,1,3-trimethyl-3-cyclohexene-5-one, 2-cyclohexen-1-one, 3,5,5-trimethyl, 3,5,5-trimethyl-2-cyclohexen-1-one, alpha-isophorone, isoacetophorone, isoforon, isoforone, isooctopherone, isophorone, izoforon | |
HJOVHMDZYOCNQW-UHFFFAOYSA-N | |
3,5,5-trimethylcyclohex-2-en-1-one | |
6544 | |
138.21 | |
97% |
Chemical Identifiers
78-59-1 | |
138.21 | |
HJOVHMDZYOCNQW-UHFFFAOYSA-N | |
6544 | |
3,5,5-trimethylcyclohex-2-en-1-one |
C9H14O | |
MFCD00001584 | |
1,1,3-trimethyl-3-cyclohexene-5-one, 2-cyclohexen-1-one, 3,5,5-trimethyl, 3,5,5-trimethyl-2-cyclohexen-1-one, alpha-isophorone, isoacetophorone, isoforon, isoforone, isooctopherone, isophorone, izoforon | |
CHEBI:34800 | |
CC1=CC(=O)CC(C1)(C)C |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Suspected of causing cancer.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
GHS Signal Word: Warning
EINECSNumber : 201-126-0
RTECSNumber : GW7700000
TSCA : TSCA