Please login to your online account to display your discounted pricing

Alfa Aesar™ Isopentyltriphenylphosphonium bromide, 98%

$32.18 - $571.07

Chemical Identifiers

Chemical Name or Material Isopentyltriphenylphosphonium bromide
CAS 28322-40-9
Molecular Formula (CH3)2CHCH2CH2P(C6H5)3Br
Formula Weight 413.35
MDL Number MFCD00031594
Beilstein 3804221
Synonym Isoamyltriphenylphosphonium bromide; (3-Methylbutyl)triphenylphosphonium bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1132209

  SDS
alfa aesar™
A1132209
10g Each for $32.18

AAA1132218

  SDS
alfa aesar™
A1132218
50g Each for $119.15

AAA1132230

  SDS
alfa aesar™
A1132230
250g Each for $571.07
Description & Specifications

Specifications

Formula Weight 413.35
Chemical Name or Material Isopentyltriphenylphosphonium bromide
Assay 98%
Molecular Formula (CH3)2CHCH2CH2P(C6H5)3Br
MDL Number MFCD00031594
CAS 28322-40-9
Beilstein 3804221
Synonym Isoamyltriphenylphosphonium bromide; (3-Methylbutyl)triphenylphosphonium bromide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈155°C
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.