Please login to your online account to display your discounted pricing

Alfa Aesar™ Isopentylamine, 99%

Chemical Identifiers

Chemical Name or Material Isopentylamine
CAS 107-85-7
Molecular Formula (CH3)2CHCH2CH2NH2
Formula Weight 87.17
MDL Number MFCD00008203
UN Number UN1106
Beilstein 1209230
Merck Index 14,5112
Synonym Isoamylamine; 3-Methylbutylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0226014

  SDS
alfa aesar™
L0226014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 87.17
Chemical Name or Material Isopentylamine
Assay 99%
Molecular Formula (CH3)2CHCH2CH2NH2
MDL Number MFCD00008203
Beilstein 1209230
Synonym Isoamylamine; 3-Methylbutylamine
Health Hazard 1 Danger
Odor Amine-like
Health Hazard 2 GHS H Statement
H225-H301-H314-H318
Highly flammable liquid and vapor.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point −60°C
Flash Point −1°C (30°F)
Quantity 25g
TSCA TSCA
CAS 107-85-7
UN Number UN1106
Merck Index 14,5112
Boiling Point 95° to 97°C
pH 12.1
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYLAMINES
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H301-H314-H318
Highly flammable liquid and vapor.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.