Please login to your online account to display your discounted pricing

Alfa Aesar™ Isopentyl mercaptan, 97%

Chemical Identifiers

Chemical Name or Material Isopentyl mercaptan
CAS 541-31-1
Molecular Formula (CH3)2CH(CH2)2SH
Formula Weight 104.22
MDL Number MFCD00004899
UN Number UN1111
Beilstein 635659
Synonym Isoamyl mercaptan; 3-Methylbutanethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL09661AC

  SDS
alfa aesar™
L09661AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL09661AE

  SDS
alfa aesar™
L09661AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 104.22
Chemical Name or Material Isopentyl mercaptan
MDL Number MFCD00004899
CAS 541-31-1
Health Hazard 3 GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay 97%
Molecular Formula (CH3)2CH(CH2)2SH
UN Number UN1111
Beilstein 635659
Synonym Isoamyl mercaptan; 3-Methylbutanethiol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H301-H332
Highly flammable liquid and vapor.
Toxic if swallowed.
Harmful if inhaled.
Flash Point 19°C (66°F)
Quantity 25mL
Boiling Point 117° to 118°C
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYL MERCAPTAN
Safety and Handling

Danger

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H301-H332
Highly flammable liquid and vapor.
Toxic if swallowed.
Harmful if inhaled.