missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Isopentyl mercaptan, 97%

Manufacturer:  Alfa Aesar™ L09661AE

 View more versions of this product

Catalog No. AAL09661AE



Specifications

Specifications

Isopentyl mercaptan
117°C to 118°C
541-31-1
MFCD00004899
635659
GIJGXNFNUUFEGH-UHFFFAOYSA-N
3-methylbutane-1-thiol
10925
97%
19°C (66°F)
100mL
C5H12S
UN1111
isoamyl mercaptan, 3-methyl-1-butanethiol, 1-butanethiol, 3-methyl, isopentyl mercaptan, 3-methylbutanethiol, isoamylthiol, isopentanethiol, isoamylmercaptan, unii-mmk4sun45e, isoamyl sulfhydrate
CC(C)CCS
104.211
104.22

Chemical Identifiers

541-31-1
104.211
GIJGXNFNUUFEGH-UHFFFAOYSA-N
10925
CC(C)CCS
C5H12S
MFCD00004899
isoamyl mercaptan, 3-methyl-1-butanethiol, 1-butanethiol, 3-methyl, isopentyl mercaptan, 3-methylbutanethiol, isoamylthiol, isopentanethiol, isoamylmercaptan, unii-mmk4sun45e, isoamyl sulfhydrate
3-methylbutane-1-thiol
Safety and Handling

Safety and Handling

GHS H Statement
H225-H301-H332
Highly flammable liquid and vapor.
Toxic if swallowed.
Harmful if inhaled.

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYL MERCAPTAN

TSCA : TSCA

Documents