Please login to your online account to display your discounted pricing

Alfa Aesar™ Isopentyl isobutyrate, 98%

Chemical Identifiers

Chemical Name or Material Isopentyl isobutyrate
CAS 2050-01-3
Molecular Formula C9H18O2
Linear Formula (CH3)2CHCH2CH2O2CCH(CH3)2
Formula Weight 158.24
MDL Number MFCD00053719
UN Number UN2620
Synonym Isobutyric acid isoamyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2487914

 
alfa aesar™
B2487914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2487922

 
alfa aesar™
B2487922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2487936

 
alfa aesar™
B2487936
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 158.24
Chemical Name or Material Isopentyl isobutyrate
TSCA TSCA
Assay 98%
Molecular Formula C9H18O2
Linear Formula (CH3)2CHCH2CH2O2CCH(CH3)2
MDL Number MFCD00053719
CAS 2050-01-3
UN Number UN2620
Synonym Isobutyric acid isoamyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 162°C
Flash Point 56°C (133°F)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMYL BUTYRATES
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.