Please login to your online account to display your discounted pricing

Alfa Aesar™ Isopentyl 3-(methylthio)propionate, 98%

$277.07 - $277.07

Chemical Identifiers

Chemical Name or Material Isopentyl 3-(methylthio)propionate
CAS 93762-35-7
Molecular Formula CH3SCH2CH2CO2CH2CH2CH(CH3)2
Formula Weight 190.31
MDL Number MFCD00053101
Beilstein 7914388
Synonym Isoamyl 3-(methylthio)propionate; 3-Methylbutyl 3-(methylthio)propionate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2231322

 
alfa aesar™
B2231322
100g Each for $277.07
Description & Specifications

Specifications

Formula Weight 190.31
Chemical Name or Material Isopentyl 3-(methylthio)propionate
Assay 98%
Molecular Formula CH3SCH2CH2CO2CH2CH2CH(CH3)2
MDL Number MFCD00053101
CAS 93762-35-7
Beilstein 7914388
Synonym Isoamyl 3-(methylthio)propionate; 3-Methylbutyl 3-(methylthio)propionate
Boiling Point 222°C
Flash Point 115°C (239°F)
Quantity 100g