missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isooctane, Uvasol™, MilliporeSigma™

For spectroscopy Uvasol™

Manufacturer:  MilliporeSigma 1047180500

 View more versions of this product

Catalog No. M1047180500



Specifications

Specifications

Isooctane
-12°C (+10°F)
Glass bottle
99°C
-107°C
540-84-1
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
NHTMVDHEPJAVLT-UHFFFAOYSA-N
2,2,4-trimethylpentane
10907
114.23
0.69g/cm3 (20°C)
410°C (770°F)
51 hPa (20°C)
Colorless
500mL
C8H18
Water: 0.56mg/L at 25°C soluble
CC(C)CC(C)(C)C
114.232
CHEBI:62805
Liquid

Chemical Identifiers

540-84-1
114.232
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
CHEBI:62805
CC(C)CC(C)(C)C
C8H18
NHTMVDHEPJAVLT-UHFFFAOYSA-N
10907
2,2,4-trimethylpentane
Safety and Handling

Safety and Handling

H304: May be fatal if swallowed and enters airways.

H315: Causes skin irritation.

H225: Highly flammable liquid and vapor

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 208-759-1

RTECSNumber : SA3320000

Recommended Storage : Keep away from heat and sources of ignition. Keep container tightly closed in a dry and well-ventilated place.

Documents
Promotions

Promotions