Please login to your online account to display your discounted pricing

Isooctane, SupraSolv™, EMD Millipore

For gas chromatography ECD and FID SupraSolv™

$208.66 - $433.86

Chemical Identifiers

Chemical Name or Material Isooctane
CAS 540-84-1
Physical Form Liquid
Molecular Formula C8H18
Formula Weight 114.23
Synonym 2,2,4-Trimethylpentane
Color Colorless
UN Number UN 1262
Melting Point −107°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1154401000

 
emd millipore corporation
1154401000
Glass bottle 1L Each for $208.66

M1154402500

 
emd millipore corporation
1154402500
Glass bottle 2.5L Each for $433.86
Description & Specifications

Specifications

Boiling Point 99°C
Formula Weight 114.23
Melting Point −107°C
Quantity 1L
Packaging Glass bottle
Health Hazard 3 H315: Causes skin irritation.
UN Number UN 1262
EINECS Number 208-759-1
Additional Information SupraSolv™ solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results
Synonym 2,2,4-Trimethylpentane
Solubility Information Water: 0.56mg/L at 25°C soluble
Density 0.69g/cm3 (20°C)
Physical Form Liquid
Health Hazard 2 H304: May be fatal if swallowed and enters airways.
Ignition Point 410°C (770°F)
DOT Information DOT Class: 3, Packaging Group: II
RTECS Number SA3320000
Color Colorless
CAS 540-84-1
Chemical Name or Material Isooctane
Molecular Formula C8H18
Vapor Pressure 51 hPa (20°C)
Flash Point -12°C (+10°F)
Health Hazard 1 H225: Highly flammable liquid and vapor
Recommended Storage Keep away from heat and sources of ignition. Keep container tightly closed in a dry and well-ventilated place.
Safety and Handling

H225: Highly flammable liquid and vapor

H315: Causes skin irritation.

H304: May be fatal if swallowed and enters airways.

Keep away from heat and sources of ignition. Keep container tightly closed in a dry and well-ventilated place.