missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isooctadecenylsuccinic Anhydride (mixture of branched chain isomers), TCI America™

$88.61 - $603.48

Chemical Identifiers

CAS 58239-72-8
Molecular Formula C22H38O3
Molecular Weight (g/mol) 350.54
MDL Number MFCD00070628
InChI Key SJSHEXCFGWSYAD-VURMDHGXNA-N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I0237500G View Documents TCI America
I0237500G
500g Each for $603.48
 
I023725G View Documents TCI America
I023725G
25g Each for $88.61
 
Specifications

Chemical Identifiers

58239-72-8
350.54
SJSHEXCFGWSYAD-VURMDHGXNA-N
C22H38O3
MFCD00070628

Specifications

Yellow
58239-72-8
MFCD00070628
350.54
Clear Liquid at 20°C
Isooctadecenylsuccinic Anhydride (mixture of branched chain isomers)
500g
C22H38O3
SJSHEXCFGWSYAD-VURMDHGXNA-N
350.54
≥90.0% (T)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0852

TSCA : Yes