Please login to your online account to display your discounted pricing

Alfa Aesar™ Isonitrosoacetophenone, 97%

$88.15 - $331.49

Chemical Identifiers

Chemical Name or Material Isonitrosoacetophenone
CAS 532-54-7
Molecular Formula C8H7NO2
Formula Weight 149.15
MDL Number MFCD00002122
Beilstein 2041691
Merck Index 14,5191
Synonym Phenylglyoxaldehyde oxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0209809

  SDS
alfa aesar™
L0209809
10g Each for $88.15

AAL0209818

  SDS
alfa aesar™
L0209818
50g Each for $331.49
Description & Specifications

Specifications

Formula Weight 149.15
Assay 97%
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Chemical Name or Material Isonitrosoacetophenone
Synonym Phenylglyoxaldehyde oxime
Molecular Formula C8H7NO2
MDL Number MFCD00002122
CAS 532-54-7
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Recommended Storage 2° to 8°C
Melting Point 124° to 128°C
Merck Index 14,5191
Quantity 10g
Beilstein 2041691
TSCA TSCA
RTECS Number MD3325000
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.

2° to 8°C