Please login to your online account to display your discounted pricing

Isonipecotic Acid Hydrochloride 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  124080250

 View more versions of this product

Catalog No. AC124080250

  • / Each
 

Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Water 0.5% max. (K.F.)
Color White to cream
Quantity 25g
Chemical Name or Material Isonipecotic acid hydrochloride
Name Note 97%
CAS 5984-56-5
Physical Form White to cream crystalline powder
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C6H11NO2·HCl
Formula Weight 165.62g/mol
MDL Number MFCD00044736
Merck Index 14, 5189
Synonym 4-Piperidinecarboxylic acid, hydrochloride
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Isonipecotic acid hydrochloride
Name Note 97%
CAS 5984-56-5
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C6H11NO2·HCl
Formula Weight 165.62g/mol
MDL Number MFCD00044736
Synonym 4-Piperidinecarboxylic acid, hydrochloride
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents