missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Isonipecotic acid, 98%

Manufacturer:  Alfa Aesar™ A1179522

 View more versions of this product

Catalog No. AAA1179522


 

Specifications

Specifications

Isonipecotic acid
498-94-2
C6H11NO2
112553
H-DL-Inp-OH; Piperidine-4-carboxylic acid
C1CNCCC1C(=O)O
Odorless
3773
>350°C
100g
98%
MFCD00006004
14,5189
SRJOCJYGOFTFLH-UHFFFAOYSA-N
piperidine-4-carboxylic acid
129.159
129.16

Chemical Identifiers

498-94-2
129.159
SRJOCJYGOFTFLH-UHFFFAOYSA-N
3773
C6H11NO2
MFCD00006004
H-DL-Inp-OH; Piperidine-4-carboxylic acid
Safety and Handling

Safety and Handling

RTECSNumber : NS5150000

TSCA : TSCA

SDS Documents