missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isonipecotamide 98.0+%, TCI America™

Manufacturer:  Tci America I0255250G

 View more versions of this product

Catalog No. I0255250G



Specifications

Specifications

Isonipecotamide
150°C
39546-32-2
MFCD00038012
DPBWFNDFMCCGGJ-UHFFFAOYSA-N
piperidine-4-carboxamide
3772
Crystal-Powder at 20°C
White
250g
C6H12N2O
4-carbamoylpiperidine, 4-piperdinecarboxamide, 4-piperidinecarboxamide, hexahydroisonicotinamide, iso nipecotamide, iso-nipecotamide, isonipecotamide, isonipectoamide, piperidin-4-carboxamid, piperidine-4-carboxylic acid amide
C1CNCCC1C(=O)N
128.175
128.18
≥98.0% (GC)

Chemical Identifiers

39546-32-2
128.175
DPBWFNDFMCCGGJ-UHFFFAOYSA-N
3772
C1CNCCC1C(=O)N
C6H12N2O
MFCD00038012
4-carbamoylpiperidine, 4-piperdinecarboxamide, 4-piperidinecarboxamide, hexahydroisonicotinamide, iso nipecotamide, iso-nipecotamide, isonipecotamide, isonipectoamide, piperidin-4-carboxamid, piperidine-4-carboxylic acid amide
piperidine-4-carboxamide
Safety and Handling

Safety and Handling

TSCA : Yes

Documents