missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isonicotinic Acid N-Oxide, TCI America™

$183.34 - $1,035.86

Chemical Identifiers

CAS 13602-12-5
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00006209
InChI Key QCWTWMJMLSKQCJ-UHFFFAOYSA-N
Synonym isonicotinic acid n-oxide, 4-carboxypyridine 1-oxide, pyridine-4-carboxylic acid n-oxide, 4-pyridinecarboxylic acid 1-oxide, 4-pyridinecarboxylic acid, 1-oxide, pyridine-4-carboxylic acid 1-oxide, 4-carboxypyridin-1-ium-1-olate, isonicotinic acid 1-oxide, 1-hydroxypyridine-4-carboxylic acid, pyridin-n-oxide-4-carboxylic acid
PubChem CID 26130
IUPAC Name 1-oxidopyridin-1-ium-4-carboxylic acid
SMILES C1=C[N+](=CC=C1C(=O)O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I0500100G View Documents TCI America
I0500100G
100g Each for $594.31
 
I050025G View Documents TCI America
I050025G
25g Each for $183.34
 
I0500250G View Documents TCI America
I0500250G
250g Each for $1,035.86
 
Specifications

Chemical Identifiers

13602-12-5
139.11
QCWTWMJMLSKQCJ-UHFFFAOYSA-N
26130
C1=C[N+](=CC=C1C(=O)O)[O-]
C6H5NO3
MFCD00006209
isonicotinic acid n-oxide, 4-carboxypyridine 1-oxide, pyridine-4-carboxylic acid n-oxide, 4-pyridinecarboxylic acid 1-oxide, 4-pyridinecarboxylic acid, 1-oxide, pyridine-4-carboxylic acid 1-oxide, 4-carboxypyridin-1-ium-1-olate, isonicotinic acid 1-oxide, 1-hydroxypyridine-4-carboxylic acid, pyridin-n-oxide-4-carboxylic acid
1-oxidopyridin-1-ium-4-carboxylic acid

Specifications

Undesignated
100g
C6H5NO3
isonicotinic acid n-oxide, 4-carboxypyridine 1-oxide, pyridine-4-carboxylic acid n-oxide, 4-pyridinecarboxylic acid 1-oxide, 4-pyridinecarboxylic acid, 1-oxide, pyridine-4-carboxylic acid 1-oxide, 4-carboxypyridin-1-ium-1-olate, isonicotinic acid 1-oxide, 1-hydroxypyridine-4-carboxylic acid, pyridin-n-oxide-4-carboxylic acid
C1=C[N+](=CC=C1C(=O)O)[O-]
139.11
139.11
≥98.0% (GC,T)
270°C
13602-12-5
MFCD00006209
QCWTWMJMLSKQCJ-UHFFFAOYSA-N
1-oxidopyridin-1-ium-4-carboxylic acid
26130
Crystal-Powder at 20°C
Isonicotinic Acid N-Oxide
Safety and Handling

Safety and Handling

TSCA : Yes