missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isonicotinic Acid N-Oxide 98.0+%, TCI America™
Supplier: TCI America I0500100G
Specifications
Isonicotinic Acid N-Oxide | |
270°C | |
MFCD00006209 | |
isonicotinic acid n-oxide, 4-carboxypyridine 1-oxide, pyridine-4-carboxylic acid n-oxide, 4-pyridinecarboxylic acid 1-oxide, 4-pyridinecarboxylic acid, 1-oxide, pyridine-4-carboxylic acid 1-oxide, 4-carboxypyridin-1-ium-1-olate, isonicotinic acid 1-oxide, 1-hydroxypyridine-4-carboxylic acid, pyridin-n-oxide-4-carboxylic acid | |
OC(=O)C1=CC=[N+]([O-])C=C1 | |
139.11 | |
139.11 | |
Crystalline Powder |
13602-12-5 | |
C6H5NO3 | |
100 g | |
QCWTWMJMLSKQCJ-UHFFFAOYSA-N | |
4-carboxypyridin-1-ium-1-olate | |
26130 | |
≥98.0% (GC,T) |
Chemical Identifiers
13602-12-5 | |
139.11 | |
QCWTWMJMLSKQCJ-UHFFFAOYSA-N | |
26130 | |
OC(=O)C1=CC=[N+]([O-])C=C1 |
C6H5NO3 | |
MFCD00006209 | |
isonicotinic acid n-oxide, 4-carboxypyridine 1-oxide, pyridine-4-carboxylic acid n-oxide, 4-pyridinecarboxylic acid 1-oxide, 4-pyridinecarboxylic acid, 1-oxide, pyridine-4-carboxylic acid 1-oxide, 4-carboxypyridin-1-ium-1-olate, isonicotinic acid 1-oxide, 1-hydroxypyridine-4-carboxylic acid, pyridin-n-oxide-4-carboxylic acid | |
4-carboxypyridin-1-ium-1-olate |
Safety and Handling
TSCA : Yes