Please login to your online account to display your discounted pricing

Alfa Aesar™ Isonicotinic acid N-oxide, 99%

$39.78 - $131.39

Chemical Identifiers

Chemical Name or Material Isonicotinic acid N-oxide
CAS 13602-12-5
Molecular Formula C6H5NO3
Formula Weight 139.11
MDL Number MFCD00006209
Beilstein 119571
Synonym Pyridine-4-carboxylic acid N-oxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0242509

  SDS
alfa aesar™
L0242509
10g Each for $39.78

AAL0242518

  SDS
alfa aesar™
L0242518
50g Each for $131.39
Description & Specifications

Specifications

Formula Weight 139.11
Assay 99%
Chemical Name or Material Isonicotinic acid N-oxide
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 13602-12-5
Synonym Pyridine-4-carboxylic acid N-oxide
MDL Number MFCD00006209
Melting Point 268° to 271°C
Quantity 10g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Beilstein 119571
TSCA TSCA
Molecular Formula C6H5NO3
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.