missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isonicotinic acid hydrazide, 99%, ACROS Organics™

$46.54 - $46.54

Chemical Identifiers

CAS 54-85-3
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
MDL Number MFCD00006426
InChI Key QRXWMOHMRWLFEY-UHFFFAOYSA-N
Synonym andrazide, isoniazid, isoniazide, isonicotinic acid hydrazide, isonicotinic hydrazide, isonicotinohydrazide, isonicotinylhydrazine, laniazid, nydrazid, rimifon
PubChem CID 3767
ChEBI CHEBI:6030
IUPAC Name pyridine-4-carbohydrazide
SMILES C1=CN=CC=C1C(=O)NN
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC122600050
SDS View Documents
Fisher Scientific Acros
122600050
5g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC122601000
SDS View Documents
Fisher Scientific Acros
122601000
100g Plastic bottle
Each for $46.54
 
AC122605000
SDS View Documents
Thermo Fisher Scientific
122605000
500g Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

54-85-3
137.14
QRXWMOHMRWLFEY-UHFFFAOYSA-N
3767
pyridine-4-carbohydrazide
C6H7N3O
MFCD00006426
andrazide, isoniazid, isoniazide, isonicotinic acid hydrazide, isonicotinic hydrazide, isonicotinohydrazide, isonicotinylhydrazine, laniazid, nydrazid, rimifon
CHEBI:6030
C1=CN=CC=C1C(=O)NN

Specifications

Authentic
Glass Bottle
171°C to 173°C
54-85-3
C6H7N3O
22, I, 504
andrazide, isoniazid, isoniazide, isonicotinic acid hydrazide, isonicotinic hydrazide, isonicotinohydrazide, isonicotinylhydrazine, laniazid, nydrazid, rimifon
QRXWMOHMRWLFEY-UHFFFAOYSA-N
pyridine-4-carbohydrazide
3767
137.14
99%
0.5% max. (1 g, 105°C)
Colorless or White
5g
99%
MFCD00006426
15, 5232
Solubility in water: 14% in water (25 c). Other solubilities: 2% in alcohol (25 c),0.1% in chloroform
C1=CN=CC=C1C(=O)NN
137.14
CHEBI:6030
Crystals or Crystalline Powder
Isonicotinic acid hydrazide, 99%
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 200-214-6

RTECSNumber : NS1751850

TSCA : TSCA

SDS
Product Certifications