Please login to your online account to display your discounted pricing

Isonicotinamide, 99%, ACROS Organics™

$23.01 - $23.01

Chemical Identifiers

Chemical Name or Material Isonicotinamide
CAS 1453-82-3
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C6H6N2O
Formula Weight 122.13g/mol
MDL Number MFCD00006432
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC122580250

  SDS
fisher scientific acros
122580250
25g Each for $23.01
Description & Specifications

Specifications

Infrared Spectrum Authentic
Color White
Melting Point 155° to 158°C
Quantity 25g
Chemical Name or Material Isonicotinamide
CAS 1453-82-3
Physical Form White crystalline powder
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C6H6N2O
Formula Weight 122.13g/mol
MDL Number MFCD00006432
Beilstein 22, 46
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin
Safety and Handling

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.