missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isoindoline hydrochloride, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC435850250
Specifications
Isoindoline hydrochloride | |
Authentic | |
White | |
25g | |
96% min. (HPLC) | |
1h-isoindole, 2,3-dihydro-, hydrochloride, 2,3-dihydro-1h-isoindole hcl, 2,3-dihydro-1h-isoindole hydrochloride, 2,3-dihydroisoindole hydrochloride, d0y1rn, dihydroisoindole hydrochloride, isoindoline hcl, isoindoline hydrochloride, isoindolinehydrochloride, ksc491i3f | |
C1C2=CC=CC=C2CN1.Cl | |
155.63 | |
155.63 | |
97% |
59.27 to 64.21% C, 8.64 to 9.36% N | |
Glass bottle | |
255°C to 256°C | |
32372-82-0 | |
C8H9N·ClH | |
NOVIRODZMIZUPA-UHFFFAOYSA-N | |
2,3-dihydro-1H-isoindole;hydrochloride | |
12311031 | |
Powder |
Chemical Identifiers
32372-82-0 | |
155.63 | |
1h-isoindole, 2,3-dihydro-, hydrochloride, 2,3-dihydro-1h-isoindole hcl, 2,3-dihydro-1h-isoindole hydrochloride, 2,3-dihydroisoindole hydrochloride, d0y1rn, dihydroisoindole hydrochloride, isoindoline hcl, isoindoline hydrochloride, isoindolinehydrochloride, ksc491i3f | |
2,3-dihydro-1H-isoindole;hydrochloride |
C8H9N·ClH | |
NOVIRODZMIZUPA-UHFFFAOYSA-N | |
12311031 | |
C1C2=CC=CC=C2CN1.Cl |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning