Please login to your online account to display your discounted pricing

Alfa Aesar™ Isoindoline hydrochloride, 95%

Chemical Identifiers

Chemical Name or Material Isoindoline hydrochloride
CAS 32372-82-0
Formula Weight 155.62
MDL Number MFCD03502377
Synonym 2,3-Dihydroisoindole hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6301203

 
alfa aesar™
H6301203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6301206

 
alfa aesar™
H6301206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63012MD

 
alfa aesar™
H63012MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 155.62
Melting Point 255° to 256°C
Chemical Name or Material Isoindoline hydrochloride
Physical Form Solid
Assay 95%
MDL Number MFCD03502377
CAS 32372-82-0
Synonym 2,3-Dihydroisoindole hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 -30° to -10°C
Sensitivity Hygroscopic
Quantity 1g

Applications
Isoindoline hydrochloride is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under inder gas. Store away aeay from oxidizing agents, water/ moisture, heat.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

-30° to -10°C