missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isoindoline, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC426280050

 View more versions of this product

Catalog No. AC426280050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Isoindoline
94°C
Glass bottle
1.05
White
5g
96% min. (GC)
MFCD00605324
GWVMLCQWXVFZCN-UHFFFAOYSA-N
2,3-dihydro-1H-isoindole
422478
Crystalline Powder, Crystals or Flakes, Darkening On Exposure To Light
1.0500g/mL
Authentic
1.5670 to 1.5700 (20°C, 589nm)
221°C
17°C
496-12-8
C8H9N
1,2-dihydroisoindole, 1,3-dihydroisoindole, 1h-isoindole, 2,3-dihydro, 2-azaindan, acmc-1am9e, dihydroisoindole, iso-indoline, isoindoline, pubchem16248
C1C2=CC=CC=C2CN1
119.17
119.17
97%

Chemical Identifiers

496-12-8
119.17
GWVMLCQWXVFZCN-UHFFFAOYSA-N
422478
C1C2=CC=CC=C2CN1
C8H9N
MFCD00605324
1,2-dihydroisoindole, 1,3-dihydroisoindole, 1h-isoindole, 2,3-dihydro, 2-azaindan, acmc-1am9e, dihydroisoindole, iso-indoline, isoindoline, pubchem16248
2,3-dihydro-1H-isoindole
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents