Please login to your online account to display your discounted pricing

Isoindoline, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC426280010

 View more versions of this product

Catalog No. AC426280010

  • / Each
 

Description & Specifications

Chemical Identifiers

Color White to off-white
Physical Form Crystalline Powder, Crystals or Flakes, Darkening On Exposure To Light
Assay ≥96.0% (GC)
Formula Weight 119.17
Quantity 1g
Density 1.0500g/mL
Infrared Spectrum Authentic
Refractive Index 1.5670 to 1.5700 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Molecular Formula C8H9N
CAS 496-12-8
Assay Percent Range 97%
Packaging Glass bottle
Chemical Name or Material Isoindoline
MDL Number MFCD00605324

Specifications

Chemical Name or Material Isoindoline
CAS 496-12-8
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C8H9N
Formula Weight 119.17
MDL Number MFCD00605324
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents