Learn More
Isohexane, for HPLC, contains <5% n-Hexane, Thermo Scientific™
Chemical Identifiers
CAS | 73513-42-5 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.18 |
MDL Number | MFCD00009406 |
InChI Key | AFABGHUZZDYHJO-UHFFFAOYSA-N |
Synonym | isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane |
PubChem CID | 7892 |
IUPAC Name | 2-methylpentane |
SMILES | CCCC(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC383810010
|
Thermo Scientific Chemicals
383810010 |
1L |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC383810025
|
Thermo Scientific Chemicals
383810025 |
2.5L |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
73513-42-5 | |
86.18 | |
AFABGHUZZDYHJO-UHFFFAOYSA-N | |
7892 | |
CCCC(C)C |
C6H14 | |
MFCD00009406 | |
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
2-methylpentane |
Specifications
73513-42-5 | |
60.0°C | |
C6H14 | |
1.3700 to 1.3720 | |
1L | |
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
AFABGHUZZDYHJO-UHFFFAOYSA-N | |
2-methylpentane | |
7892 | |
HPLC |
0.6600g/mL | |
−23°C | |
CH3CH2CH2CH(CH3)2 | |
MFCD00009406 | |
0.66 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol | |
CCCC(C)C | |
86.18 | |
86.18 | |
Isohexane |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapour.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Danger
RUO â Research Use Only