Learn More
Isoeugenol, 98+%, mixture of cis/trans isomers, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 122575000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Isoeugenol | |
97-54-1 | |
Yellow | |
266.0°C to 268.0°C | |
Authentic | |
Glass bottle | |
1.5760 to 1.5780 | |
MFCD00009285 | |
06,II,916 | |
15, 5216 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether and other organic,solvents | |
COC1=CC(\C=C\C)=CC=C1O | |
164.20 | |
26.7 mPa.s (25°C) | |
164.2 | |
Viscous Liquid |
99% | |
19.0°C to 23.0°C | |
1.0900g/mL | |
150°C | |
98% min. (GC) | |
C10H12O2 | |
CH3OC6H3(CH=CHCH3)OH | |
500 g | |
1.09 | |
isoeugenol, e-isoeugenol, trans-isoeugenol, 2-methoxy-4-propenylphenol, 4-propenylguaiacol, 4-hydroxy-3-methoxy-1-propenylbenzene, 2-methoxy-4-prop-1-en-1-yl phenol, trans-p-propenylquaiacol, isoeugenol trans-form, 4-hydroxy-3-methoxypropenylbenzene | |
BJIOGJUNALELMI-ONEGZZNKSA-N | |
2-methoxy-4-[(E)-prop-1-enyl]phenol | |
853433 | |
CHEBI:50545 | |
98+% |
Chemical Identifiers
97-54-1 | |
164.20 | |
BJIOGJUNALELMI-ONEGZZNKSA-N | |
853433 | |
COC1=CC(\C=C\C)=CC=C1O |
C10H12O2 | |
MFCD00009285 | |
isoeugenol, e-isoeugenol, trans-isoeugenol, 2-methoxy-4-propenylphenol, 4-propenylguaiacol, 4-hydroxy-3-methoxy-1-propenylbenzene, 2-methoxy-4-prop-1-en-1-yl phenol, trans-p-propenylquaiacol, isoeugenol trans-form, 4-hydroxy-3-methoxypropenylbenzene | |
CHEBI:50545 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse
GHS Signal Word: Warning
EINECSNumber : 202-590-7
RTECSNumber : SL7875000
TSCA : TSCA
RUO – Research Use Only