Please login to your online account to display your discounted pricing

Isocytosine, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398320050

 View more versions of this product

Catalog No. AC398320050

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 111.1
Assay ≥98.5%
Physical Form Crystalline Powder
Quantity 5g
Color White to off-white
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material Isocytosine
CAS 108-53-2
MDL Number MFCD01109044
Assay Percent Range 99%
Molecular Formula C4H5N3O
Synonym 2-Amino-4-hydroxypyrimidine

Specifications

Chemical Name or Material Isocytosine
CAS 108-53-2
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C4H5N3O
Formula Weight 111.1
MDL Number MFCD01109044
Synonym 2-Amino-4-hydroxypyrimidine
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents