missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isoconazole Nitrate 98.0+%, TCI America™

$229.09 - $709.89

Chemical Identifiers

CAS 24168-96-5
Molecular Formula C18H15Cl4N3O4
Molecular Weight (g/mol) 479.135
MDL Number MFCD01721647
InChI Key NNGQLSIGRSTLLU-UHFFFAOYSA-N
Synonym 1-2,4-dichloro-beta-2,6-dichlorobenzyloxy phenethyl imidazole nitrate, 1-2-2,6-dichlorobenzyl oxy-2-2,4-dichlorophenyl ethyl imidazole nitrate, adestan g100, fazol, isoconazole nitrate, isoconazole nitrate jan, isoconazole; nitric acid, travogyn
PubChem CID 159968
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(2,6-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid
SMILES C1=CC(=C(C(=C1)Cl)COC(CN2C=CN=C2)C3=C(C=C(C=C3)Cl)Cl)Cl.[N+](=O)(O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I08345G View Documents Tci America
I08345G
5g Each for $229.09
 
I083425G View Documents Tci America
I083425G
25g Each for $709.89
 
Specifications

Chemical Identifiers

24168-96-5
479.135
NNGQLSIGRSTLLU-UHFFFAOYSA-N
159968
C1=CC(=C(C(=C1)Cl)COC(CN2C=CN=C2)C3=C(C=C(C=C3)Cl)Cl)Cl.[N+](=O)(O)[O-]
C18H15Cl4N3O4
MFCD01721647
1-2,4-dichloro-beta-2,6-dichlorobenzyloxy phenethyl imidazole nitrate, 1-2-2,6-dichlorobenzyl oxy-2-2,4-dichlorophenyl ethyl imidazole nitrate, adestan g100, fazol, isoconazole nitrate, isoconazole nitrate jan, isoconazole; nitric acid, travogyn
1-[2-(2,4-dichlorophenyl)-2-[(2,6-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid

Specifications

White-Yellow
5g
C18H15Cl4N3O4
1-2,4-dichloro-beta-2,6-dichlorobenzyloxy phenethyl imidazole nitrate, 1-2-2,6-dichlorobenzyl oxy-2-2,4-dichlorophenyl ethyl imidazole nitrate, adestan g100, fazol, isoconazole nitrate, isoconazole nitrate jan, isoconazole; nitric acid, travogyn
C1=CC(=C(C(=C1)Cl)COC(CN2C=CN=C2)C3=C(C=C(C=C3)Cl)Cl)Cl.[N+](=O)(O)[O-]
479.135
479.14
≥98.0% (HPLC,T)
183°C
24168-96-5
MFCD01721647
NNGQLSIGRSTLLU-UHFFFAOYSA-N
1-[2-(2,4-dichlorophenyl)-2-[(2,6-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid
159968
Crystal-Powder at 20°C
Isoconazole Nitrate
Safety and Handling

Safety and Handling

RTECSNumber : NI4772000

TSCA : No