Please login to your online account to display your discounted pricing

Isochroman 99%, ACROS Organics™

$40.97 - $104.24

Chemical Identifiers

Chemical Name or Material Isochroman
Name Note 99%
CAS 493-05-0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H10O
Formula Weight 134.18
MDL Number MFCD00006913
Synonym 3, 4-Dihydro-1H-2-benzopyran
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC122540250

  SDS
thermo fisher scientific
122540250
Glass bottle 25g Each for $104.24

AC122540050

  SDS
thermo fisher scientific
122540050
Glass bottle 5g Each for $40.97
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 134.18
Quantity 25g
Color Clear colorless to yellow
Beilstein 17, 53
EINECS Number 207-774-0
Density 1.06g/mL
Infrared Spectrum Authentic
Refractive Index 1.5430 to 1.5460 (20°C, 589nm)
Packaging Glass bottle
Synonym 3, 4-Dihydro-1H-2-benzopyran
MDL Number MFCD00006913
Name Note 99%
CAS 493-05-0
Chemical Name or Material Isochroman
Assay Percent Range 99%
Molecular Formula C9H10O