Please login to your online account to display your discounted pricing

Alfa Aesar™ Isochroman, 99%

$53.73 - $183.18

Chemical Identifiers

Chemical Name or Material Isochroman
CAS 493-05-0
Molecular Formula C9H10O
Formula Weight 134.18
MDL Number MFCD00006913
Synonym 3,4-Dihydro-1H-2-benzopyran
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1256609

 
alfa aesar™
A1256609
10g Each for $53.73

AAA1256618

 
alfa aesar™
A1256618
50g Each for $183.18
Description & Specifications

Specifications

Formula Weight 134.18
Molecular Formula C9H10O
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Chemical Name or Material Isochroman
Assay 99%
MDL Number MFCD00006913
CAS 493-05-0
Synonym 3,4-Dihydro-1H-2-benzopyran
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 89° to 90°C (12mmHg)
Flash Point 66°C (151°F)
Quantity 10g

Applications
Isochroman is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.